• USA Home
  • Search Results

  • 2ethyl3methoxypyrazine138172568058411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Ethyl-3-methoxypyrazine

2 Products
Image of 2-<WBR>Ethyl-<WBR>3-<WBR>methoxypyrazine
CAS Number:
Empirical Formula (Hill Notation):
 C7H10N2O
Molecular Weight:
 138.17
 EC Number:
 FEMA Number:
 3280
 Flavis number:
 14.112
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More