• USA Home
  • Search Results

  • 2ethylbutylacetate144211003187511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Ethylbutyl acetate

1 Products
Image of 2-Ethylbutyl acetate
CAS Number:
Linear Formula:
 CH3CO2CH2CH(C2H5)2
Molecular Weight:
 144.21
 Council of Europe no.:
 215
 EC Number:
 FEMA Number:
 2425
 Flavis number:
 9.025
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More