• USA Home
  • Search Results

  • 2ethylhexyl4dimethylaminobenzoate277402124502311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Ethylhexyl 4-(dimethylamino)benzoate

1 Products
Synonym:   2-Ethylhexyl 4-(N ,N -dimethylamino)benzoate, 2-Ethylhexyl 4-(dimethylamino)benzoate, 2-Ethylhexyl N ,N -dimethyl-p -aminobenzoate, 2-Ethylhexyl p -(dimethylamino)benzoate, 4-(Dimethylamino)benzoic acid 2-ethylhexyl ester, EHDAB, Octyl dimethyl PABA
Image of 2-Ethylhexyl 4-<WBR>(dimethylamino)<WBR>benzoate
CAS Number:
Linear Formula:
 (CH3)2NC6H4CO2CH2CH(C2H5)(CH2)3CH3
Molecular Weight:
 277.40
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More