• USA Home
  • Search Results

  • 2ethylhexylacrylate1842810311711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Ethylhexyl acrylate

3 Products
Synonym:   (±)-Acrylic acid 2-ethylhexyl ester
Image of 2-Ethylhexyl acrylate
CAS Number:
Linear Formula:
 CH2=CHCOOCH2CH(C2H5)(CH2)3CH3
Molecular Weight:
 184.28
 Beilstein/REAXYS Number:
 1765828
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Additive
  Application(s)
  Potency
  Density
Show More