• USA Home
  • Search Results

  • 2ethylpyrazine108141392500311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Ethylpyrazine

1 Products
Synonym:   Ethylpyrazine
Image of 2-Ethylpyrazine
CAS Number:
Empirical Formula (Hill Notation):
 C6H8N2
Molecular Weight:
 108.14
 Beilstein/REAXYS Number:
 108200
 Council of Europe no.:
 2213
 EC Number:
 FEMA Number:
 3281
 Flavis number:
 14.022
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More