• USA Home
  • Search Results

  • 2fluoro4nitrotoluene15513142707211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Fluoro-4-nitrotoluene

1 Products
Image of 2-<WBR>Fluoro-<WBR>4-<WBR>nitrotoluene
CAS Number:
Linear Formula:
 CH3C6H3(NO2)F
Molecular Weight:
 155.13
 Beilstein/REAXYS Number:
 2250156
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More