• USA Home
  • Search Results

  • 2fluorophenylacetonitrile1351432662511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Fluorophenylacetonitrile

1 Products
Synonym:   2-Fluorobenzyl cyanide
Image of 2-<WBR>Fluorophenylacetonitrile
CAS Number:
Linear Formula:
 FC6H4CH2CN
Molecular Weight:
 135.14
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More