• USA Home
  • Search Results

  • 2furanmethanethiol114179802211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Furanmethanethiol

2 Products
Synonym:   2-Furfurylthiol, 2-Furylmethanethiol, Furfuryl mercaptan
Image of 2-Furanmethanethiol
CAS Number:
Empirical Formula (Hill Notation):
 C5H6OS
Molecular Weight:
 114.17
 Beilstein/REAXYS Number:
 383594
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More