• USA Home
  • Search Results

  • 2heptanone1141911043011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Heptanone

5 Products
Synonym:   Methyl amyl ketone, n-Amyl methyl ketone, Methyl pentyl ketone
Image of 2-Heptanone
CAS Number:
Linear Formula:
 CH3(CH2)4COCH3
Molecular Weight:
 114.19
 EC Index Number
 203-767-1
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Assay
  Special Grade
Show More