• USA Home
  • Search Results

  • 2hexadecanone240421878763811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Hexadecanone

1 Products
Synonym:   Methyl tetradecyl ketone
Image of 2-Hexadecanone
CAS Number:
Linear Formula:
 CH3(CH2)13COCH3
Molecular Weight:
 240.42
 Beilstein/REAXYS Number:
 1769889
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
374180 2-Hexadecanone, 98%,