• USA Home
  • Search Results

  • 2hydroxy4methoxybenzophenone2282413157711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Hydroxy-4-methoxybenzophenone

1 Products
Synonym:   Oxybenzone
Image of 2-<WBR>Hydroxy-<WBR>4-<WBR>methoxybenzophenone
CAS Number:
Linear Formula:
 HOC6H3(OCH3)COC6H5
Molecular Weight:
 228.24
 Beilstein/REAXYS Number:
 1913145
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More