• USA Home
  • Search Results

  • 2hydroxy4methyl3nitropyridine154122190118811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Hydroxy-4-methyl-3-nitropyridine

1 Products
Synonym:   4-Methyl-3-nitro-2-pyridone
Image of 2-<WBR>Hydroxy-<WBR>4-<WBR>methyl-<WBR>3-<WBR>nitropyridine
CAS Number:
Empirical Formula (Hill Notation):
 C6H6N2O3
Molecular Weight:
 154.12
 Beilstein/REAXYS Number:
 139125
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More