• USA Home
  • Search Results

  • 2hydroxy4nitrobenzoicacid1831261919211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Hydroxy-4-nitrobenzoic acid

1 Products
Image of 2-<WBR>Hydroxy-<WBR>4-<WBR>nitrobenzoic acid
CAS Number:
Linear Formula:
 HOC6H3(NO2)CO2H
Molecular Weight:
 183.12
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More