• USA Home
  • Search Results

  • 2hydroxydodecanoicacid21632298455611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Hydroxydodecanoic acid

1 Products
Synonym:   α-Hydroxylauric acid, 2-Hydroxylauric acid, DL-α-Hydroxylauric acid
Image of 2-Hydroxydodecanoic acid
CAS Number:
Linear Formula:
 CH3(CH2)9CH(OH)COOH
Molecular Weight:
 216.32
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More