• USA Home
  • Search Results

  • 2hydroxyethylacrylate1161281861111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Hydroxyethyl acrylate

1 Products
Synonym:   Ethylene glycol monoacrylate
Image of 2-Hydroxyethyl acrylate
CAS Number:
Linear Formula:
 CH2=CHCOOCH2CH2OH
Molecular Weight:
 116.12
 Beilstein/REAXYS Number:
 969853
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More