• USA Home
  • Search Results

  • 2hydroxyoctanoicacid1602161773211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-2-Hydroxyoctanoic acid

1 Products
Synonym:   (±)-2-Hydroxycaprylic acid, 2-Hydroxycaprylic acid
Image of (±)<WBR>-<WBR>2-<WBR>Hydroxyoctanoic acid
CAS Number:
Linear Formula:
 CH3(CH2)5CH(OH)COOH
Molecular Weight:
 160.21
 Beilstein/REAXYS Number:
 1760638
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More