• USA Home
  • Search Results

  • 2hydroxyquinoline4carboxylicacid189178490681011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Hydroxyquinoline-4-carboxylic acid

1 Products
Image of 2-<WBR>Hydroxyquinoline-<WBR>4-<WBR>carboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C10H7NO3
Molecular Weight:
 189.17
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More