• USA Home
  • Search Results

  • 2imidazolidone860912093411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Imidazolidone

2 Products
Synonym:   2-Oxoimidazolidine, N,N′-Ethyleneurea
Image of 2-Imidazolidone
CAS Number:
Empirical Formula (Hill Notation):
 C3H6N2O
Molecular Weight:
 86.09
 Beilstein/REAXYS Number:
 106252
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More