• USA Home
  • Search Results

  • 2iodoanisole2340352928211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Iodoanisole

1 Products
Synonym:   2-Iodophenyl methyl ether, 2-Methoxyiodobenzene
Image of 2-Iodoanisole
CAS Number:
Linear Formula:
 IC6H4OCH3
Molecular Weight:
 234.03
 Beilstein/REAXYS Number:
 1860243
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More

Product #
Description
252786 2-Iodoanisole, 98%,