• USA Home
  • Search Results

  • 2iodohippuricacid3050714758011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Iodohippuric acid

1 Products
Synonym:   N-(2-Iodobenzoyl)glycine
Image of 2-Iodohippuric acid
CAS Number:
Linear Formula:
 IC6H4CONHCH2CO2H
Molecular Weight:
 305.07
 Beilstein/REAXYS Number:
 1877943
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
106577 2-Iodohippuric acid, 98%,