• USA Home
  • Search Results

  • 2iodothiophene21004343795411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Iodothiophene

1 Products
Image of 2-Iodothiophene
CAS Number:
Empirical Formula (Hill Notation):
 C4H3IS
Molecular Weight:
 210.04
 Beilstein/REAXYS Number:
 104666
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
196150 2-Iodothiophene, 98%,