• USA Home
  • Search Results

  • 2isobutyl3methoxypyrazine166222468300911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Isobutyl-3-methoxypyrazine

3 Products
Image of 2-<WBR>Isobutyl-<WBR>3-<WBR>methoxypyrazine
CAS Number:
Empirical Formula (Hill Notation):
 C9H14N2O
Molecular Weight:
 166.22
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More