• USA Home
  • Search Results

  • 2isopropenylaniline133195256219311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Isopropenylaniline

1 Products
Synonym:   2-(1-Methylethenyl)aniline, 2-(1-Methylethenyl)benzenamine, 2-(Prop-1-en-2-yl)aniline, o -Isopropenylaniline
Image of 2-Isopropenylaniline
CAS Number:
Linear Formula:
 H2C=C(CH3)C6H4NH2
Molecular Weight:
 133.19
 Beilstein/REAXYS Number:
 3081217
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More