• USA Home
  • Search Results

  • 2isopropylphenol136198869711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Isopropylphenol

3 Products
Synonym:   2-(1-Methylethyl)phenol, 2-Hydroxycumene
Image of 2-Isopropylphenol
CAS Number:
Empirical Formula (Hill Notation):
 C9H12O
Molecular Weight:
 136.19
 EC Index Number
 201-852-8
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More