• USA Home
  • Search Results

  • 2mercaptobenzoxazole15119238296911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Mercaptobenzoxazole

1 Products
Synonym:   2-Benzoxazolethiol
Image of 2-<WBR>Mercaptobenzoxazole
CAS Number:
Empirical Formula (Hill Notation):
 C7H5NOS
Molecular Weight:
 151.19
 Beilstein/REAXYS Number:
 119483
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
M3507 2-Mercaptobenzoxazole, 95%,