• USA Home
  • Search Results

  • 2mercaptoethanol78136024211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Mercaptoethanol

6 Products
Synonym:   Thioethylene glycol, Thioglycol
Image of 2-Mercaptoethanol
CAS Number:
Empirical Formula (Hill Notation):
 C2H6OS
Molecular Weight:
 78.13
 EC Index Number
 200-464-6
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  UV-Absorption Values
  Restriction
  Product Line
  Solubility
  Special Grade
Show More