• USA Home
  • Search Results

  • 2methoxy3methylpyrazine12414284730511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methoxy-3-methylpyrazine

2 Products
Image of 2-<WBR>Methoxy-<WBR>3-<WBR>methylpyrazine
CAS Number:
Empirical Formula (Hill Notation):
 C6H8N2O
Molecular Weight:
 124.14
 Council of Europe no.:
 2266
 EC Number:
 FEMA Number:
 3183
 Flavis number:
 14.126
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More