• USA Home
  • Search Results

  • 2methoxybenzaldehyde1361513502411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methoxybenzaldehyde

2 Products
Synonym:   o-Anisaldehyde, Salicylaldehyde methyl ether
Image of 2-<WBR>Methoxybenzaldehyde
CAS Number:
Linear Formula:
 CH3OC6H4CHO
Molecular Weight:
 136.15
 Beilstein/REAXYS Number:
 606301
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
  Potency
Show More