• USA Home
  • Search Results

  • 2methoxycinnamaldehyde16219150474111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methoxycinnamaldehyde

2 Products
Synonym:   o-Methoxycinnamaldehyde
Image of 2-<WBR>Methoxycinnamaldehyde
CAS Number:
Linear Formula:
 CH3OC6H4CH=CHCHO
Molecular Weight:
 162.19
 Council of Europe no.:
 571
 EC Number:
 FEMA Number:
 3181
 Flavis number:
 5.048
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More