• USA Home
  • Search Results

  • 2methoxyethanol760910986411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methoxyethanol

7 Products
Synonym:   Ethylene glycol monomethyl ether, Methyl glycol
Image of 2-Methoxyethanol
CAS Number:
Linear Formula:
 CH3OCH2CH2OH
Molecular Weight:
 76.09
 Beilstein/REAXYS Number:
 1731074
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Density
  Color
  Restriction
  Refractive Index
  Additive
  UV-Absorption Values
  Application(s)
  pH Value
  Physical Form
  Special Grade
Show More