• USA Home
  • Search Results

  • 2methoxyethylacetate1181311049611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methoxyethyl acetate

3 Products
Synonym:   1-Acetoxy-2-methoxyethane, Ethylene glycol monomethyl ether acetate, Methyl Cellosolve® acetate
Image of 2-Methoxyethyl acetate
CAS Number:
Linear Formula:
 CH3COOCH2CH2OCH3
Molecular Weight:
 118.13
 Beilstein/REAXYS Number:
 1700761
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  Application(s)
  Potency
  Physical Form
  Explosion limit
  Assay
Show More