• USA Home
  • Search Results

  • 2methyl1butanol881513732611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-2-Methyl-1-butanol

6 Products
Synonym:   sec-Butyl carbinol
Image of (±)<WBR>-<WBR>2-<WBR>Methyl-<WBR>1-<WBR>butanol
CAS Number:
Linear Formula:
 CH3CH2CH(CH3)CH2OH
Molecular Weight:
 88.15
 Beilstein/REAXYS Number:
 1839383
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Refractive Index
  Document Links
  Special Grade
  Allergen
  Organoleptic property
  Vapor pressure
  Application(s)
  Potency
  Solubility
  Assay
Show More