• USA Home
  • Search Results

  • 2methyl1butene701356346211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methyl-1-butene

2 Products
Image of 2-Methyl-1-butene
CAS Number:
Linear Formula:
 CH3CH2C(CH3)=CH2
Molecular Weight:
 70.13
 Beilstein/REAXYS Number:
 505975
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More