• USA Home
  • Search Results

  • 2methyl1phenyl1propene1322076849011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methyl-1-phenyl-1-propene

1 Products
Synonym:   (2-Methylpropenyl)benzene
Image of 2-<WBR>Methyl-<WBR>1-<WBR>phenyl-<WBR>1-<WBR>propene
CAS Number:
Linear Formula:
 C6H5CH=C(CH3)2
Molecular Weight:
 132.20
 Beilstein/REAXYS Number:
 1902046
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More