• USA Home
  • Search Results

  • 2methyl1propanol74127883111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methyl-1-propanol

10 Products
Synonym:   Isobutanol, Isobutyl alcohol
Image of 2-Methyl-1-propanol
CAS Number:
Linear Formula:
 (CH3)2CHCH2OH
Molecular Weight:
 74.12
 Beilstein/REAXYS Number:
 1730878
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  UV-Absorption Condition
  Purity Grade
  Product Line
  Application(s)
  Refractive Index
  UV-Absorption Values
  Physical Form
  Assay
Show More