• USA Home
  • Search Results

  • 2methyl2butanol88157585411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methyl-2-butanol

4 Products
Synonym:   tert-Amyl alcohol, tert-Pentyl alcohol
Image of 2-Methyl-2-butanol
CAS Number:
Linear Formula:
 CH3CH2C(CH3)2OH
Molecular Weight:
 88.15
 Beilstein/REAXYS Number:
 1361351
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  pH Value
  Vapor pressure
  Application(s)
  Product Line
  Physical Form
Show More