• USA Home
  • Search Results

  • 2methyl2butene701351335911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methyl-2-butene

4 Products
Synonym:   β-Isoamylene, Amylene
Image of 2-Methyl-2-butene
CAS Number:
Linear Formula:
 CH3CH=C(CH3)2
Molecular Weight:
 70.13
 Beilstein Registry Number:
 1361353
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Titertest
  Refractive Index
  Vapor pressure
Show More