• USA Home
  • Search Results

  • 2methyl3buten2ol861311518411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methyl-3-buten-2-ol

3 Products
Synonym:   1,1-Dimethylallyl alcohol, 3-Hydroxy-3-methyl-1-butene
Image of 2-<WBR>Methyl-<WBR>3-<WBR>buten-<WBR>2-<WBR>ol
CAS Number:
Linear Formula:
 CH2=CHC(CH3)2OH
Molecular Weight:
 86.13
 Beilstein/REAXYS Number:
 1698263
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More