• USA Home
  • Search Results

  • 2methyl3butyn2ol841211519511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methyl-3-butyn-2-ol

1 Products
Synonym:   Dimethyl ethynyl carbinol
Image of 2-<WBR>Methyl-<WBR>3-<WBR>butyn-<WBR>2-<WBR>ol
CAS Number:
Linear Formula:
 HC≡CC(CH3)2OH
Molecular Weight:
 84.12
 Beilstein/REAXYS Number:
 635746
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More