• USA Home
  • Search Results

  • 2methyl4isothiazolin3onehydrochloride151612617254311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methyl-4-isothiazolin-3-one hydrochloride

1 Products
Synonym:   MIT, Methylisothiazolinone hydrochloride
Image of 2-<WBR>Methyl-<WBR>4-<WBR>isothiazolin-<WBR>3-<WBR>one hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C4H5NOS · HCl
Molecular Weight:
 151.61
 Beilstein Registry Number:
 3993054
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More