• USA Home
  • Search Results

  • 2methyl4nitroaniline152159952511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methyl-4-nitroaniline

1 Products
Synonym:   2-Amino-5-nitrotoluene, 4-Nitro-o-toluidine
Image of 2-<WBR>Methyl-<WBR>4-<WBR>nitroaniline
CAS Number:
Linear Formula:
 CH3C6H3(NO2)NH2
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 775772
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More