• USA Home
  • Search Results

  • 2methylallylaminehydrochloride107582814854111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methylallylamine hydrochloride

1 Products
Synonym:   2-Methyl-2-propen-1-amine hydrochloride
Image of 2-Methylallylamine hydrochloride
CAS Number:
Linear Formula:
 H2C=C(CH3)CH2NH2 · HCl
Molecular Weight:
 107.58
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More