• USA Home
  • Search Results

  • 2methylbutane72157878411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methylbutane

7 Products
Synonym:   Isopentane
Image of 2-Methylbutane
CAS Number:
Linear Formula:
 CH3CH2CH(CH3)2
Molecular Weight:
 72.15
 Beilstein/REAXYS Number:
 1730723
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Refractive Index
  UV-Absorption Condition
  Product Line
  Special Grade
  Application(s)
  UV-Absorption Values
  Melting Point
  Physical Form
Show More