• USA Home
  • Search Results

  • 2methylbutyraldehyde86139617311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methylbutyraldehyde

3 Products
Synonym:   2-Methylbutanal
Image of 2-<WBR>Methylbutyraldehyde
CAS Number:
Linear Formula:
 C2H5CH(CH3)CHO
Molecular Weight:
 86.13
 Beilstein/REAXYS Number:
 1633540
 Council of Europe no.:
 575
 EC Number:
 FEMA Number:
 2691
 Flavis number:
 5.049
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Special Grade
  Assay
Show More