• USA Home
  • Search Results

  • 2methylcyclohexanone1121758360811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methylcyclohexanone

3 Products
Synonym:   Tetrahydro-o-cresol
Image of 2-<WBR>Methylcyclohexanone
CAS Number:
Linear Formula:
 CH3C6H9(=O)
Molecular Weight:
 112.17
 Beilstein/REAXYS Number:
 506751
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
Show More