• USA Home
  • Search Results

  • 2methylindole131179520511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methylindole

2 Products
Synonym:   NSC 7514
Image of 2-Methylindole
CAS Number:
Empirical Formula (Hill Notation):
 C9H9N
Molecular Weight:
 131.17
 Beilstein/REAXYS Number:
 109781
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More