• USA Home
  • Search Results

  • 2methylisoborneolsolution16828237142811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methylisoborneol solution

3 Products
Synonym:   (1R-exo)-1,2,7,7-Tetramethylbicyclo[2.2.1]heptan-2-ol
Image of 2-Methylisoborneol solution
CAS Number:
Empirical Formula (Hill Notation):
 C11H20O
Molecular Weight:
 168.28
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Special Grade
  Titertest
  Package quantity
Show More