• USA Home
  • Search Results

  • 2methylpyrazine941110908011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methylpyrazine

4 Products
Image of 2-Methylpyrazine
CAS Number:
Empirical Formula (Hill Notation):
 C5H6N2
Molecular Weight:
 94.11
 Beilstein/REAXYS Number:
 105778
 Council of Europe no.:
 2270
 EC Number:
 FEMA Number:
 3309
 Flavis number:
 14.027
 MDL number:
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Anal. Standard Form
  Physical Form
  Assay
  Special Grade
Show More