• USA Home
  • Search Results

  • 2methylresorcinol1241460825311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Methylresorcinol

2 Products
Synonym:   2,6-Dihydroxytoluene
Image of 2-Methylresorcinol
CAS Number:
Linear Formula:
 CH3C6H3(OH)2
Molecular Weight:
 124.14
 Beilstein/REAXYS Number:
 2042177
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More