• USA Home
  • Search Results

  • 2naphthaldehyde156186699911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Naphthaldehyde

1 Products
Synonym:   β-Naphthaldehyde
Image of 2-Naphthaldehyde
CAS Number:
Linear Formula:
 C10H7CHO
Molecular Weight:
 156.18
 Beilstein/REAXYS Number:
 507750
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
N206 2-Naphthaldehyde, 98%,